missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Borosilicate Glass Narrow Neck Erlenmeyer Flask

Used for a wide range of laboratory experiments. Pyrex™ Borosilicate Glass Narrow Neck Erlenmeyer Flask is DIN ISO 1773 compliant.

Brand:  Pyrex 1130/26D

149.28 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11922619

  • £259.00 / Pack of 10
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Used for a wide range of laboratory experiments. Pyrex™ Borosilicate Glass Narrow Neck Erlenmeyer Flask is DIN ISO 1773 compliant.

  • Complies with ISO 1773 and DIN 12353
  • Erlenmeyer pattern
  • Manufactured from Pyrex™ borosilicate glass for highest resistance to thermal shock and chemical corrosion
  • White enamel graduations and marking spot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ISO 1773, DIN 12353
Borosilicate Glass
220 mm
Yes
131 mm
1000 mL
Yes
Conical
Yes
35
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers