missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Büchner Flask with Screw Thread Connector

Manufactured from borosilicate glass. Pyrex™ Büchner Flask with Screw Thread Connector features heavy wall construction for vacuum filtration applications, and tooled neck to accept a drilled rubber stopper. Sidearm includes a PTFE screwthread connector for easy and safe fitting of 9mm bore vacuum tubing. Complies with ISO 6556.

Brand:  Pyrex 1170/06SC

385.78 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.



Code : A0

Additional Details : Weight : 4.86900kg

 View more versions of this product

Product Code. 11337834

  • £585.00 / Pack of 10
Estimated Shipment: 23-10-2024
to see stock.
Add to basket

Description

Description

Manufactured from borosilicate glass. Pyrex™ Büchner Flask with Screw Thread Connector features heavy wall construction for vacuum filtration applications, and tooled neck to accept a drilled rubber stopper. Sidearm includes a PTFE screwthread connector for easy and safe fitting of 9mm bore vacuum tubing. Complies with ISO 6556.

  • Complies with ISO 6556
  • Büchner flasks with heavy wall construction for vacuum filtration applications
  • sidearm features a PTFE screwthread connector for easy and safe fitting of 9mm bore vacuum tubing
  • With tooled neck to accept a drilled rubber stopper
Specifications

Specifications

ISO 6556, DIN 12476
500 mL
185 mm
105 mm
35 mm (Neck I.D.)
Borosilicate Glass
PTFE screw thread connector
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers