missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Büchner Flask with Screw Thread Connector

Manufactured from borosilicate glass. Pyrex™ Büchner Flask with Screw Thread Connector features heavy wall construction for vacuum filtration applications, and tooled neck to accept a drilled rubber stopper. Sidearm includes a PTFE screwthread connector for easy and safe fitting of 9mm bore vacuum tubing. Complies with ISO 6556.

Brand:  Pyrex 1170/08SC

290.17 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.



Code : A0

Additional Details : Weight : 4.75000kg

 View more versions of this product

Product Code. 12559125

  • £437.00 / Pack of 5
Estimated Shipment: 20-08-2024
to see stock.
Add to basket

Description

Description

Manufactured from borosilicate glass. Pyrex™ Büchner Flask with Screw Thread Connector features heavy wall construction for vacuum filtration applications, and tooled neck to accept a drilled rubber stopper. Sidearm includes a PTFE screwthread connector for easy and safe fitting of 9mm bore vacuum tubing. Complies with ISO 6556.

  • Complies with ISO 6556
  • Büchner flasks with heavy wall construction for vacuum filtration applications
  • sidearm features a PTFE screwthread connector for easy and safe fitting of 9mm bore vacuum tubing
  • With tooled neck to accept a drilled rubber stopper
Specifications

Specifications

ISO 6556, DIN 12476
1000 mL
230 mm
135 mm
45 mm (Neck I.D.)
Borosilicate Glass
PTFE screw thread connector
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers