missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PYREX™ Cloning Cylinders
GREENER_CHOICE

Create identical cells by surrounding a single cell or group of cells

Brand:  Corning™ 3166-10

 View more versions of this product

Product Code. 11997595

  • £78.19 / Pack of 125
Estimated Shipment: 07-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Clone a single cell or group of cells by surrounding them with this glass cylinder. Dip the end of the cylinder into a sterile silicone grease before pressing to the bottom of a culture flask to create an ­isolated well.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Borosilicate Glass
10 mm
0.39 in.
0.39 in.
10 mm
Non-sterile
Cloning Cylinder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ PYREX™ Cloning Cylinders > 10 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.