missing translation for 'onlineSavingsMsg'
Learn More

PYREX™ Borosilicate Glass Graduated Cylinders

Printed with blue enamel graduations for enhanced visibility. Pyrex™ Borosilicate Glass Cylinders in Tall Form provide superior chemical resistance.

Brand:  Pyrex 3280-1L

 View more versions of this product

Product Code. 12682042

  • £213.00 / Pack of 2
Estimated Shipment: 30-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Printed with blue enamel graduations for enhanced visibility. Pyrex™ Borosilicate Glass Cylinders in Tall Form provide superior chemical resistance.

  • Hexagonal base delivers stability
  • Heavy duty construction increases mechanical strength and reduces breakage (lowering cost and improving user safety)
  • Heavily beaded rim and spout allow clean and easy dispensing
  • Class B
  • Complies with ISO 4788
Complies: ISO 4788
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hexagonal
Round
±10 mL
BS 604, DIN 12680, ISO 4788
1000 mL
Blue enamel
Blue
Graduated Cylinder
Borosilicate Glass
B
465 mm
10 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers