Learn More
Corning™ PYREX™ Heavy-Duty Griffin Beakers
GREENER_CHOICE
For use under harsh conditions such as mechanized washing
£183.00 - £937.00
Specifications
Material Borosilicate Glass
Type Beaker
Graduated Yes
Style Griffin
View More Specs
Products 9
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
12549995
GREENER_CHOICE
View Documents Promotion Details
Corning™
1003-150
150 mL
£222.00
Pack of 12
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
12559995
View Documents Promotion Details
Corning™
1003-250
250 mL
£281.00
Pack of 12
In Stock
to see stock.
Edge
Add to basket
12569995
View Documents Promotion Details
Corning™
1003-400
400 mL
£301.00
Pack of 12
In Stock
to see stock.
Edge
Add to basket
12671766
View Documents Promotion Details
Corning™
1003-600
600 mL
£183.00
Pack of 6
In Stock
to see stock.
Edge
Add to basket
12509975
View Documents Promotion Details
Corning™
1003-1L
1 L
£277.00
Pack of 6
In Stock
to see stock.
Edge
Add to basket
11911516
View Documents Promotion Details
Corning™
1003-2L
2000 mL
£326.00
Pack of 4
In Stock
to see stock.
Edge
Add to basket
10343010
View Documents Promotion Details
Corning™
1003-3L
3000 mL
£343.00
Each
Estimated Shipment: 15-05-2025
to see stock.
Ajouter au panier
10337879
missing translation for 'viewDocuments' Détails de la promotion
Corning™
1003-4L
4000 mL
£191.00
Each
Estimated Shipment: 22-05-2025
to see stock.
Ajouter au panier
10829610
missing translation for 'viewDocuments' Détails de la promotion
Corning™
1003-5L
5000 mL
£937.00
Each
Estimated Shipment: 15-05-2025
to see stock.
Ajouter au panier
Description
Description
Measure liquids with sturdy, long-lasting, reinforced glass. Pyrex™ Borosilicate Glass Heavy-duty Griffin Beakers are available in a wide range of sizes
Measure liquids with sturdy, long-lasting, reinforced glass. Pyrex™ Borosilicate Glass Heavy-duty Griffin Beakers are available in a wide range of sizes.
Heavy duty beakers specifically designed to offer the best mechanical strength under harsh conditions, such as mechanized washing operations. For convenience, the 250 through 4000 mL beakers have a double graduated metric scale to indicate their approximate content. All sizes have an extra large marking spot.
Compliance
Designed to ASTM™ E 960 requirements.
Spécification
Spécification
Borosilicate Glass
Beaker
Yes
Griffin
Vidéos
FDS
Documents
Documents
Certificats
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Corning™ PYREX™ Heavy-Duty Griffin Beakers
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis