Learn More
Pyrex™ Scoop with Hollow Stem
Aids the introduction of powder into narrow neck vessels. Pyrex™ Scoop with Hollow Stem sits flat to ensure stability on the balance pan. Available in 3 capacities.
£344.00 - £446.00
Specifications
Material Borosilicate Glass
Product Type Weighing scoop
View More Specs
Products 3
Product Code Brand Capacity (Metric) Height (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Height (Metric) Price Quantity & Availability  
12651992
View Documents Promotion Details
Pyrex
2115/06M
10 mL 100 mm
£446.00
Pack of 10
Estimated Shipment: 29-05-2025
to see stock.
Add to basket
12631992
View Documents Promotion Details
Pyrex
2115/02M
3 mL 70 mm
£344.00
Pack of 10
In Stock
to see stock.
Edge
Add to basket
11970885
View Documents Promotion Details
Pyrex
2115/04M
6 mL 85 mm
£367.00
Pack of 10
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
Description
Description
Aids the introduction of powder into narrow neck vessels. Pyrex™ Scoop with Hollow Stem sits flat to ensure stability on the balance pan. Available in 3 capacities.
  • With hollow stem to aid introduction of the powder into narrow neck vessels like volumetric flasks
  • The flat base ensures stability on the balance pan
Specifications
Specifications
Borosilicate Glass
Weighing scoop
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Pyrex™ Scoop with Hollow Stem