missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine, 99+%, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Metal Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -42.0°C |
Color | Colorless |
Density | 0.9780g/mL |
Boiling Point | 115.0°C to 116.0°C |
Quantity | 500 mL |
Flash Point | 17°C |
Packaging | EcoSafPak™ Glass Bottle |
Physical Form | Liquid |
Refractive Index | 1.5085 to 1.5105 |
Assay Percent Range | 99+% |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0458110 | Certificate of Analysis | 30/01/2025 | 418545000,418540250,418540010,418540025 |
A0461351 | Certificate of Analysis | 25/03/2024 | 418540250,418545000,418540010,418540025 |
A0458110 | Certificate of Analysis | 23/11/2023 | 418540250,418545000,418540010,418540025 |
B0541438B | Certificate of Analysis | 14/11/2023 | 418540250,418545000,418540010,418540025 |
B0539219A | Certificate of Analysis | 20/10/2023 | 418540250,418545000,418540010,418540025 |
Safety and Handling


Product Identifier
- Pyridine
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P240-Ground/bond container and receiving equipment.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Pyridine, 99+%, ACS reagent, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction