missing translation for 'onlineSavingsMsg'
Learn More
Pyridine, 99+%, for spectroscopy, Thermo Scientific Chemicals Product Code.: 10215752

Pyridine, 99+%, for spectroscopy, Thermo Scientific Chemicals

Product Code. 10215752
250 mL, Glass Bottle
Click to view available options
Quantity:
25 mL
250 mL
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10215752

Brand: Thermo Scientific Acros 386562500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
MDL Number MFCD00011732
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717Show MoreShow Less
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

Melting Point -42.0°C
Absorbance 0.022 max. at 345nm, 0.097 max. at 340nm, 0.301 max. at 335nm, 1 max. at 310nm
Density 0.9780g/mL
Boiling Point 115.0°C to 116.0°C
Quantity 250 mL
Flash Point 17°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.5085 to 1.5105
Assay Percent Range 99+%
Residue after Evaporation 0.001% max.
Specific Gravity 0.978
Viscosity 0.95 mPa.s (20°C)
Water 0.05% max.
Merck Index 15, 8081
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and most other common, organic solvents
Formula Weight 79.1
Percent Purity ≥99%
Grade Spectroscopy
Chemical Name or Material Pyridine, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468293 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number386562500,386560010,386560250
Lot NumberA0465298 Certificate TypeCertificate of Analysis Date14/08/2024 Catalog Number386562500,386560010,386560250
Lot NumberA0450018 Certificate TypeCertificate of Analysis Date15/02/2024 Catalog Number386562500,386560010,386560250
Lot NumberA0455840 Certificate TypeCertificate of Analysis Date31/08/2023 Catalog Number386562500,386560010,386560250
Lot NumberA0366095 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number386562500,386560010,386560250

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pyridine, 99+%, for spectroscopy, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.