missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Pyridostatin pentahydrochloride
Stabilizes G-quadruplexes; induces DNA damage and cell cycle arrest
Brand: Tocris Bioscience™ 4763/10
172.78 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Binds and stabilizes G-quadruplexes, inducing DNA damage and cell cycle arrest (Kd = 490 nM); targets the proto-oncogene Src, reducing Src protein abundance and Src-dependent motility in human breast cancer cells. Also targets telomeric G-quadruplexes, inducing telomerase dysfunction. Activates the DNA-dependent protein kinase catalytic sunbunit (DNA-PKcs).Specifications
95% | |
1781882-65-2 | |
10 mg | |
M.W. (Observed): 778.94 |
4-(2-Aminoethoxy)N2,N6-bis[(4-(2-aminoethoxy)-2-quinolinyl]-2,6-pyridinecarboxamide pentahydrochloride | |
Desiccate at RT | |
Pyridostatin pentahydrochloride | |
Pyridostatin pentahydrochloride |
For Research Use Only