Learn More
Chemical Identifiers
CAS | 109-97-7 |
---|---|
Molecular Formula | C4H5N |
Molecular Weight (g/mol) | 67.09 |
MDL Number | MFCD00005216 |
InChI Key | KAESVJOAVNADME-UHFFFAOYSA-N |
Synonym | pyrrole, divinylenimine, azole, imidole, pyrrol, monopyrrole, divinyleneimine, 1-aza-2,4-cyclopentadiene, polypyrrole, unii-86s1zd6l2c |
PubChem CID | 8027 |
ChEBI | CHEBI:19203 |
IUPAC Name | 1H-pyrrole |
SMILES | N1C=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10011840
|
Thermo Scientific Acros
|
25 g | Glass bottle |
£26.33
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10498310
|
Thermo Scientific Acros
|
100 g | Glass bottle |
£96.48
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10764461
|
Thermo Scientific Acros
|
500 g | Glass bottle |
£217.08
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11926611
|
Thermo Scientific Acros
|
2.5 kg | Glass bottle |
£846.21
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
109-97-7 | |
67.09 | |
KAESVJOAVNADME-UHFFFAOYSA-N | |
8027 | |
1H-pyrrole |
C4H5N | |
MFCD00005216 | |
pyrrole, divinylenimine, azole, imidole, pyrrol, monopyrrole, divinyleneimine, 1-aza-2,4-cyclopentadiene, polypyrrole, unii-86s1zd6l2c | |
CHEBI:19203 | |
N1C=CC=C1 |
Specifications
109-97-7 | |
Brown to Colorless | |
129.0°C to 131.0°C | |
Authentic | |
C4H5N | |
MFCD00005216 | |
15,8128 | |
pyrrole, divinylenimine, azole, imidole, pyrrol, monopyrrole, divinyleneimine, 1-aza-2,4-cyclopentadiene, polypyrrole, unii-86s1zd6l2c | |
KAESVJOAVNADME-UHFFFAOYSA-N | |
1H-pyrrole | |
8027 | |
1.317 mPa.s (20°C) | |
99% | |
Liquid |
-23.0°C | |
0.9600g/mL | |
36°C | |
98.5% min. (GC) | |
1.5080 to 1.5100 | |
20,159 | |
0.96 | |
Solubility in water: 60g/L (20°C). Other solubilities: insoluble in dilute alkalies, soluble in alcohol, ether and dilute acids, soluble in most organic solvents | |
N1C=CC=C1 | |
67.09 | |
CHEBI:19203 | |
67.09 | |
Extra Pure | |
Pyrrole |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Causes serious eye damage.
Toxic if swallowed.
Flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-724-7
missing translation for 'rtecsNumber' : UX9275000
missing translation for 'tsca' : TSCA
Recommended Storage : Darkens on storage
- Pyrrole
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
Hazard Statement
- H226-Flammable liquid and vapour.
- H301-Toxic if swallowed.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.