missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Q-BioTaq™ DNA Polymerase Multiplex PCR Kit
A thermostable DNA polymerase which can be used effectively for multiplex PCR.
Brand: MP Biomedicals™ 11EPQBT015
Code : 50
Additional Details : Weight : 1.00000kg
Description
- Used to amplify DNA sequences
- It is particularly useful in applications where multiple targets are amplified (multiplex)
- Contains all the reagents (presented as separate items) required for PCR including: Q-BioTaq™ DNA polymerase, high purity dNTPs Mix (>99%), optimized incubation buffers with and without MgCl2 and a separate MgCl2.
- Research Use Only (RUO)
Specifications
5 U/μL | |
Kit | |
Molecular Biology, PCR/Amplification, PCR, cDNA synthesis, RT-PCR, DNA labeling, Multiplex PCR, RAPD, Random Amplified Polymorphic DNA | |
Multiplex PCR Kit | |
No |
Store at -20°C | |
Any DNA Isolation or Purification Kit | |
Q-BioTaq™ DNA Polymerase, 10x Q-BioTaq™ Buffer with Mg, 10x Q-BioTaq™ Buffer without Mg, 25 mM MgCl2 solution | |
150 U |