missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Q-BioTaq™ DNA Polymerase Multiplex PCR Kit

A thermostable DNA polymerase which can be used effectively for multiplex PCR.

£73.56 - £391.14

Specifications

Concentration 5 U/μL
Content And Storage Store at -20°C
Format Kit
For Use With (Equipment) Any DNA Isolation or Purification Kit
For Use With (Application) Molecular Biology, PCR/Amplification, PCR, cDNA synthesis, RT-PCR, DNA labeling, Multiplex PCR, RAPD, Random Amplified Polymorphic DNA
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11440466
View Documents
MP Biomedicals™
11EPQBT010
100 U This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11467000
View Documents
MP Biomedicals™
11EPQBT015
150 U This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Used to amplify DNA sequences
  • It is particularly useful in applications where multiple targets are amplified (multiplex)
  • Contains all the reagents (presented as separate items) required for PCR including: Q-BioTaq™ DNA polymerase, high purity dNTPs Mix (>99%), optimized incubation buffers with and without MgCl2 and a separate MgCl2.
  • Research Use Only (RUO)
Specifications

Specifications

5 U/μL
Kit
Molecular Biology, PCR/Amplification, PCR, cDNA synthesis, RT-PCR, DNA labeling, Multiplex PCR, RAPD, Random Amplified Polymorphic DNA
Multiplex PCR Kit
Store at -20°C
Any DNA Isolation or Purification Kit
Q-BioTaq™ DNA Polymerase, 10x Q-BioTaq™ Buffer with Mg, 10x Q-BioTaq™ Buffer without Mg, 25 mM MgCl2 solution
No
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.