Learn More
Socorex™ Qualitix™ Sterilization Pouch
Fits any small to mid-size sterilization device and provides easy and effective wrapping of items. Made of robust medical grade paper, they tightly close with self-sealing strip to maintain sterility. Content remains visible through PET/CPP transparent laminated film.
Brand:  Socorex™ 3210.1936
 View more versions of this product
Product Code. 15740679
  • £62.50 / Pack of 200
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Robust durable material
  • Quick content identification
  • Large self-sealing strip, easy to close
  • Chevron top seal for easy opening
  • Suitable for steam and EO gas sterilization
  • Two sterilization indicators
  • Five different sizes
  • Conformity to ISO 17665-06, ISO 11135, CE
Specifications
Specifications
Autoclavable
Self-adhesive
190 x 360 mm
ISO 17665-06, ISO 11135, CE
Paper
Product Suggestions
Product Suggestions
Videos
Videos
  • Socorex Qualitix sterilisation pouches
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Socorex™ Qualitix™ Sterilization Pouch > 190 x 360 mm