Learn More
Sartorius Quantitative Grade 391 Filter Papers
Used for quantitative and gravimetric analyses, as well as for pressure or vacuum filtration. Sartorius Qualitative Grade 391 Filter Papers are made of 100% cotton linters with an α-cellulose content of >98%.
£8.20 - £248.00
Specifications
Basis Weight | 84 g/m2 |
---|---|
Color Code | Blue |
Material | Cotton Linters |
Particle Retention | 2 to 3 μm |
Filtration Speed | 180 sec. |
Product Code | Brand | Diameter (Metric) | Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Format | Price | Quantity & Availability | ||||
11741385
|
Sartorius
FT-4-104-125 |
125 mm | Folded |
£62.40
Pack of 100 |
Estimated Shipment: 10-09-2024
Log in to see stock. |
||||
15902860
|
Sartorius
FT-3-104-063 |
63 mm | Disc |
£13.10
Pack of 100 |
Estimated Shipment: 12-09-2024
Log in to see stock. |
||||
10578001
|
Sartorius
FT-3-104-125 |
125 mm | Disc |
£37.50
Pack of 100 |
In Stock
Log in to see stock. |
||||
10420283
|
Sartorius
FT-3-104-090 |
90 mm | Disc |
£23.20
Pack of 100 |
In Stock
Log in to see stock. |
||||
11761385
|
Sartorius
FT-4-104-185 |
185 mm | Folded |
£91.80
Pack of 100 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
10400093
|
Sartorius
FT-3-104-150 |
150 mm | Disc |
£53.60
Pack of 100 |
In Stock
Log in to see stock. |
||||
10664522
|
Sartorius
FT-3-104-045 |
45 mm | Disc |
£11.10
Pack of 100 |
In Stock
Log in to see stock. |
||||
15992850
|
Sartorius
FT-3-104-050 |
50 mm | Disc |
£11.10
Pack of 100 |
Estimated Shipment: 12-09-2024
Log in to see stock. |
||||
11771385
|
Sartorius
FT-4-104-240 |
240 mm | Folded |
£143.00
Pack of 100 |
Estimated Shipment: 10-09-2024
Log in to see stock. |
||||
10409902
|
Sartorius
FT-3-104-185 |
185 mm | Disc |
£80.60
Pack of 100 |
In Stock
Log in to see stock. |
||||
10325511
|
Sartorius
FT-3-104-240 |
240 mm | Disc |
£248.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
10059470
|
Sartorius
FT-3-104-070 |
70 mm | Disc |
£14.50
Pack of 100 |
In Stock
Log in to see stock. |
||||
10263811
|
Sartorius
FT-3-104-055 |
55 mm | Disc |
£13.20
Pack of 100 |
In Stock
Log in to see stock. |
||||
15972850
|
Sartorius
FT-3-104-035 |
35 mm | Disc |
£8.20
Pack of 100 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
10243861
|
Sartorius
FT-3-104-110 |
110 mm | Disc |
£32.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
15942980
|
Sartorius
FT-4-104-150 |
150 mm | Folded |
£74.70
Pack of 100 |
Estimated Shipment: 10-09-2024
Log in to see stock. |
||||
Description
- Acid-washed to achieve high purity
- Ash-free (ash content ≤ 0.01 % according to DIN 540370)
- Wet-strengthened
- Color-coded box for easy selection
Specifications
84 g/m2 | |
Cotton Linters | |
180 sec. | |
391 | |
0.15 mm |
Blue | |
2 to 3 μm | |
quantitative and gravimetric analyses, pressure or vacuum filtration | |
100/Pk. |