Learn More
Applied Biosystems™ QuantStudio 7 Flex package
Delivers the proven reliability, sensitivity, and accuracy of the Applied Biosystems ViiA 7 System in a new industrial design. The QuantStudio 7 Flex System has been optimized to enable the broadest range of quantitative PCR applications, with additional dyes, formats, and automation options.
Brand:  Applied Biosystems™ A27185
 View more versions of this product
Product Code. 15701178
  • £73025.00 / Each
Explore more special offers
Description
Description
  • Affordable price allows you to obtain a high performance instrument with minimal upfront capital investment
  • Compatible with over 8 million TaqMan™ assays for gene expression, genetic variation, gene regulation, or protein expression experiments
  • The intelligent design of the QuantStudio™ platform combined with Applied Biosystems™ instrument reliability and accuracy
  • Easy-to-use software, responsive touch-screen, and effortless block exchange without the need for any tools
  • The QuantStudio™ 6 Flex system accommodates theversatility of 4 interchangeable blocks
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PC (Laptop)
96
54.7 cm
1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance)
4°C to 99.9°C
96-well FAST, 6.5°C/s
Optiflex System Lamp
FAM™, SYBR™ Green, VIC™, JOE™, HEX™, TET™, ABY™, NED™, TAMRA™, Cy™3, JUN™, ROX™, Texas Red™, Mustang Purple™, Cy™5, LIZ™, Cy™5.5
96-well FAST
0.25°C
Touchscreen
33.8 cm
52.5 cm
QuantStudio 7 Flex
6.0°C/s (96-well FAST)
0.1 mL (96-well FAST)
38 kg