Learn More
Applied Biosystems™ QuantStudio 7 Flex package
Delivers the proven reliability, sensitivity, and accuracy of the Applied Biosystems ViiA 7 System in a new industrial design. The QuantStudio 7 Flex System has been optimized to enable the broadest range of quantitative PCR applications, with additional dyes, formats, and automation options.
£73025.00 - £99655.00
Specifications
Thermal Uniformity | 0.25°C |
---|---|
Display Type | Touchscreen |
Height (Metric) | 54.7 cm |
Width (Metric) | 33.8 cm |
Depth (Metric) | 52.5 cm |
Product Code | Brand | Content And Storage | Block Configurations | Warranty | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Content And Storage | Block Configurations | Warranty | Price | Quantity & Availability | |||
15721178
|
Applied Biosystems™
A27187 |
PC (Laptop) | 96-well | 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) |
£75325.00
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15711168
|
Applied Biosystems™
A27174 |
PC (Laptop) | 96-well | 1 Year Manufacturer's + 3 Year Field Service repair (with 1 planned maintenance/Year) |
£90755.00
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15701178
|
Applied Biosystems™
A27185 |
PC (Laptop) | 96-well FAST | 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) |
£73025.00
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15721168
|
Applied Biosystems™
A27175 |
PC (Laptop) | 96-well FAST | 1 Year Manufacturer's + 3 Year Field Service repair (with 1 planned maintenance/Year) |
£88455.00
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15741178
|
Applied Biosystems™
A27189 |
PC (Laptop) | 384-well | 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) |
£76625.00
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15791158
|
Applied Biosystems™
A27172 |
PC (Laptop) | 384-well | 1 Year Manufacturer's + 3 Year Field Service repair (with 1 planned maintenance/Year) |
£92055.00
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15761178
|
Applied Biosystems™
A27191 |
PC (Laptop) | TaqMan Array Card | 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) |
£84225.00
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15711178
|
Applied Biosystems™
A27186 |
PC (Tower) | 96-well | 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) |
£75325.00
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15741168
|
Applied Biosystems™
A27177 |
PC (Tower) | 96-well | 1 Year Manufacturer's + 3 Year Field Service repair (with 1 planned maintenance/Year) |
£90755.00
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15791168
|
Applied Biosystems™
A27184 |
PC (Tower) | 96-well FAST | 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) |
£73025.00
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15731168
|
Applied Biosystems™
A27176 |
PC (Tower) | 96-well FAST | 1 Year Manufacturer's + 3 Year Field Service repair (with 1 planned maintenance/Year) |
£88455.00
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15731178
|
Applied Biosystems™
A27188 |
PC (Tower) | 384-well | 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) |
£76625.00
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15701168
|
Applied Biosystems™
A27173 |
PC (Tower) | 384-well | 1 Year Manufacturer's + 3 Year Field Service repair (with 1 Planned Maintenance/Year) |
£92055.00
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15751178
|
Applied Biosystems™
A27190 |
PC (Tower) | TaqMan Array Card | 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) |
£84225.00
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15781158
|
Applied Biosystems™
A27171 |
PC (Tower) | TaqMan Array Card | 1 Year Manufacturer's + 3 Year Field Service repair (with 1 planned maintenance/Year) |
£99655.00
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Affordable price allows you to obtain a high performance instrument with minimal upfront capital investment
- Compatible with over 8 million TaqMan™ assays for gene expression, genetic variation, gene regulation, or protein expression experiments
- The intelligent design of the QuantStudio™ platform combined with Applied Biosystems™ instrument reliability and accuracy
- Easy-to-use software, responsive touch-screen, and effortless block exchange without the need for any tools
- The QuantStudio™ 6 Flex system accommodates theversatility of 4 interchangeable blocks
Specifications
0.25°C | |
54.7 cm | |
52.5 cm | |
QuantStudio 7 Flex | |
38 kg |
Touchscreen | |
33.8 cm | |
4°C to 99.9°C | |
FAM™, SYBR™ Green, VIC™, JOE™, HEX™, TET™, ABY™, NED™, TAMRA™, Cy™3, JUN™, ROX™, Texas Red™, Mustang Purple™, Cy™5, LIZ™, Cy™5.5 |