Learn More
Macherey-Nagel™ Quartz Wool
Brand: Macherey-Nagel™ 718587
70.35 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Specifications
25G | |
Quartz Wool | |
Quartz |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Macherey-Nagel™ Quartz Wool > For Use With: Used for GC liners, packed GC columns etc.
Spot an opportunity for improvement?Share a Content Correction