Learn More
Invitrogen™ Qubit™ 1X dsDNA High Sensitivity (HS) and Broad Range (BR) Assay Kits

Highly selective for dsDNA over RNA, the Qubit 1X dsDNA HS and BR Assay Kits offer easy-to-use formulations for the accurate quantitation of dsDNA in 100 and 500 assay formats. Lambda Standards provide sufficient material for up to 500 samples.
£120.00 - £365.00
Product Code | Brand | Quantity | Assay | Quantitation Range | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Assay | Quantitation Range | Price | Quantity & Availability | |||
15850210
![]() |
Invitrogen™
Q33230 |
100 assays | 1X dsDNA, high sensitivity | 0.1 to 120 ng |
£121.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15860210
![]() |
Invitrogen™
Q33231 |
500 assays | 1X dsDNA, high sensitivity | 0.1 to 120 ng |
£365.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17104589
![]() |
Invitrogen™
Q33266 |
500 assays | 1X dsDNA, broad range | 4 to 4000 ng |
£354.00
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17194104
![]() |
Invitrogen™
Q33265 |
100 assays | 1X dsDNA, broad range | 4 to 4000 ng |
£120.00
67g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Experience easy, accurate, and precise quantification of dsDNA with Qubit 1X dsDNA HS (High Sensitivity) and Qubit 1X dsDNA BR (Broad Range) Assay Kits. The 1X formulation provides a ready-to-use working solution that enables simple dsDNA sample quantification on Qubit Fluorometers. These assays are highly selective for dsDNA over ssDNA, RNA, protein, and free nucleotides. Contaminants, such as salts, solvents, or detergents are well-tolerated.
Qubit 1X dsDNA HS and BR Assay Kits, designed for use with Qubit Fluorometers, are highly selective for double-stranded DNA (dsDNA) over single-stranded DNA (ssDNA), RNA, protein, and free nucleotides. All kits provide a ready-to-use working solution and DNA standards. To perform the assay, simply dilute your sample (any volume from 1–20 μL is acceptable) into the 1X working solution provided, then read the concentration using a Qubit Fluorometer.Qubit 1X dsDNA HS Assay Kit
The Qubit dsDNA HS (High Sensitivity) Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of sensitive DNA samples. Depending on sample volume, the assay kit is designed to be accurate for initial DNA sample concentrations of 5 pg/μL to 120 ng/μL, providing a detection range of 0.1−120 ng.
Qubit 1X dsDNA BR Assay Kit
The Qubit dsDNA BR (Broad-Range) Assay Kit, designed for use with Qubit 4 or Qubit Flex Fluorometers, provides an accurate and selective method for the quantitation of DNA samples. Depending on sample volume, the assay kit is designed to be accurate for initial DNA sample concentrations of 0.2 to 4,000 ng/μL, providing a detection range of 4-4,000 ng.
Notes
• Qubit 1X dsDNA HS can be used with Qubit 2, Qubit 3, Qubit 4, and Qubit Flex Fluorometers
• Qubit 1X dsDNA BR are designed for use with Qubit 4 and Qubit Flex Fluorometers
• Use thin-wall, clear, 0.5-mL PCR tubes (Cat. No. Q32856) for the Qubit 4 Fluorometer and 8 x 200 μL tube strips (Cat. No. Q33252) for the Qubit Flex Fluorometer
Specifications
Fewer resources used, Less waste | |
Qubit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.