missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quickfit™ Borosilicate Glass Complete Soxhlet Extractor Assembly
Complete kit including all glass components necessary to perform a solid-liquid extraction. Quickfit™ Borosilicate Glass Complete Soxhlet Extractor Assembly includes an extractor, condenser, and an evaporating flask.
Brand: Quickfit™ 100RASX
262.43 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Each assembly comprises the basic components for solid-liquid extraction : Flask/Soxhlet extractor/Condenser/Associated lids and clips
- Choice of assemblies with different extractor capacities

Specifications
Soxhlet Extraction Assembly | |
Pyrex Borosilicate Glass | |
Ideal for solid-liquid extraction |
100 mL | |
250mL FR250/3S flask, EX5/63 Soxhlet extractor, associated lids and clips, CX7/06/SC condenser |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Quickfit™ Borosilicate Glass Complete Soxhlet Extractor Assembly > Capacity: 100mL
Spot an opportunity for improvement?Share a Content Correction