Learn More
Quickfit™ Borosilicate Glass Condenser Coil with Ground Cone
Circulates coolant through a coil with a very large cooling surface. Quickfit™ Borosilicate Glass Condenser Coil with Ground Cone is ideal for use in the upright reflux mode where counter-current cooling is possible. Supplied with detachable plastic connectors for the attachment of rubber hose.
Brand:  Quickfit CX3/05/SC
93.12 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 12060063
  • £183.00 / Each
Estimated Shipment: 28-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Circulates coolant through a coil with a very large cooling surface. Quickfit™ Borosilicate Glass Condenser Coil with Ground Cone is ideal for use in the upright reflux mode where counter-current cooling is possible. Supplied with detachable plastic connectors for the attachment of rubber hose.
  • Coolant is circulated through a coil having a very large cooling surface 
  • Ideal for use in the upright reflux mode where counter-current cooling is possible
  • Supplied with detachable plastic connectors for the attachment of rubber hose
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ideal for use in the upright reflux mode where counter-current cooling is possible
34/35
Condenser Coil
320 mm (Overall) 160 mm (Effective)
Borosilicate Glass