Learn More
Quickfit™ Borosilicate Glass Cylindrical Culture Vessel
Manufactured from temperature resistant borosilicate glass. Quickfit™ Borosilicate Glass Cylindrical Culture Vessel is ideal for fermentation and culture work. Autoclavable with precision ground flat-flange joint. For use with Quickfit™ flat-flange lids and retaining clips.
£112.00 - £3170.00
Product Code | Brand | Capacity (Metric) | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
11371995
|
Quickfit
FV1L |
1000 mL | 105 mm | 160 mm |
£112.00
Each |
Estimated Shipment: 10-03-2025
Log in to see stock. |
||||
11391995
|
Quickfit
FV2L |
2000 mL | 133 mm | 225 mm |
£318.00
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
||||
11312005
|
Quickfit
FV5L |
5000 mL | 181 mm | 290 mm |
£270.00
Each |
Estimated Shipment: 10-03-2025
Log in to see stock. |
||||
15507910
|
Quickfit
FV10L |
10,000 mL | 227 mm | 375 mm |
£2025.00
Pack of 5 |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||||
15517910
|
Quickfit
FV20L |
20,000 mL | 288 mm | 465 mm |
£3170.00
Pack of 5 |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||
15527910
|
Quickfit
FV500 |
500 mL | 80 mm | 130 mm |
£798.00
Pack of 5 |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||
Description
Manufactured from temperature resistant borosilicate glass. QuickfitTM Borosilicate Glass Cylindrical Culture Vessel is ideal for fermentation and culture work. Autoclavable with precision ground flat-flange joint. For use with QuickfitTM flat-flange lids and retaining clips.
- Ideal for fermentation and culture work
- Manufactured from temperature resistant borosilicate glass
- Autoclavable
- With precision ground flat-flange joint
- For use with Quickfit™ flat-flange lids and retaining clips
Specifications
Flat | |
Borosilicate Glass | |
1 Each |