Learn More
Quickfit™ Borosilicate Glass Drying Angled Tube
Keep material securely dry. QuickFit™ Borosilicate glass drying angled cone to fit QuickFitT™ apparatus.
Brand:  Quickfit MF23/2
34.50 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12098371
  • £36.04 / Each
Estimated Shipment: 06-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with a desiccant such as calcium chloride
  • Ground glass cone to fit Quickfit™ apparatus
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Quickfit™ apparatus
Borosilicate Glass
Tube
19/26
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Quickfit™ Borosilicate Glass Drying Angled Tube > Cone Shape: 19/26