Learn More
Quickfit™ Borosilicate Glass Drying Angled Tube
Keep material securely dry. QuickFit™ Borosilicate glass drying angled cone to fit QuickFitT™ apparatus.
£36.04 - £233.00
Specifications
Material Borosilicate Glass
Product Type Tube
For Use With (Equipment) Quickfit™ apparatus
View More Specs
Products 4
Product Code Brand Standard Taper Joint No. Price Quantity & Availability  
Product Code Brand Standard Taper Joint No. Price Quantity & Availability  
12088371
View Documents
Quickfit
MF23/1
14/23
£38.35
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
12098371
View Documents
Quickfit
MF23/2
19/26
£36.04
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
12008381
View Documents
Quickfit
MF23/3
24/29
£53.55
Each
Estimated Shipment: 15-04-2025
to see stock.
Add to basket
15527940
View Documents
Quickfit
MF23/4
29/32
£233.00
Pack of 5
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • For use with a desiccant such as calcium chloride
  • Ground glass cone to fit Quickfit™ apparatus
Specifications
Specifications
Borosilicate Glass
Tube
Quickfit™ apparatus
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Quickfit™ Borosilicate Glass Drying Angled Tube