missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quickfit™ Borosilicate Glass Jacketed Coil Condenser
Complies with DIN 12593. Quickfit™ Borosilicate Glass Jacketed Coil Condenser has an outer water jacket which provides a large cooling surface. Jacket avoids vapour creeping around the coil. Ideal for reflux work.
Brand: Quickfit CX6/33/SC
203.67 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Complies with DIN 12593
- Coil condenser has an outer water jacket
- Coil provides large cooling surface
- Jacket avoids vapour creeping around the coil
- Ideal for reflux work
Specifications
DIN 12593 | |
Ideal for reflux work | |
24/29 | |
Jacketed Coil Condenser |
Overall Length: 365mm | |
160 mm | |
Borosilicate Glass |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction