missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Borosilicate Glass Jacketed Reaction Flask

Contains an outer jacket for the circulation of a temperature controlled liquid (water, oil, etc.)

Brand:  Quickfit JRV2L

 View more versions of this product

Product Code. 15547920

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Flat flange top joint to allow easy access to the flasks contents
  • Ideal for temperature controlled reactions
  • For use with Quickfit flat-flange lids and retaining clips
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2000 mL
Ideal for temperature controlled reactions and for circulation of water or oil etc
312 mm
Cylindrical
Borosilicate Glass
Quickfit™ flat flange lids and retaining clips
Reaction flask
165 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.