missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Borosilicate Glass Jacketed Reaction Flask

Contains an outer jacket for the circulation of a temperature controlled liquid (water, oil, etc.)

£2805.00 - £3450.00

Specifications

Material Borosilicate Glass
For Use With (Application) Ideal for temperature controlled reactions and for circulation of water or oil etc
For Use With (Equipment) Quickfit™ flat flange lids and retaining clips
Product Type Reaction flask
Shape Cylindrical
View More Specs

Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15537920
View Documents
Quickfit
JRV1L
1000 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15547920
View Documents
Quickfit
JRV2L
2000 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Flat flange top joint to allow easy access to the flasks contents
  • Ideal for temperature controlled reactions
  • For use with Quickfit flat-flange lids and retaining clips
Specifications

Specifications

Borosilicate Glass
Quickfit™ flat flange lids and retaining clips
Cylindrical
Ideal for temperature controlled reactions and for circulation of water or oil etc
Reaction flask
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.