Learn More
Quickfit™ Borosilicate Glass Liebig Condenser
Popular water-jacketed condenser suitable for distillation and reflux applications. The Quickfit Corosilicate Glass Liebig Condenser hase a detachable plastic screwthread connectors for hosing.
Brand: Quickfit C1/12/SC
50.40 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Description
Popular water-jacketed condenser suitable for distillation and reflux applications. The Quickfit Corosilicate Glass Liebig Condenser hase a detachable plastic screwthread connectors for hosing.
- Complies with DIN 12576
- Popular water-jacketed condenser suitable for distillation and reflux applications
- With detachable plastic screwthread connectors for easy and safe attachment of rubber hose
Specifications
DIN 12576 | |
340 mm | |
Borosilicate Glass |
Suitable for distillation and reflux applications | |
19/26 | |
Liebig Condenser |
Product Content Correction
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido