Learn More
Quickfit™ Borosilicate Glass Liebig Condenser
Popular water-jacketed condenser suitable for distillation and reflux applications. The Quickfit Corosilicate Glass Liebig Condenser hase a detachable plastic screwthread connectors for hosing.
Brand:  Quickfit C1/12/25/SC
48.34 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12644911
  • £84.05 / Each
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Popular water-jacketed condenser suitable for distillation and reflux applications. The Quickfit Corosilicate Glass Liebig Condenser hase a detachable plastic screwthread connectors for hosing.
  • Complies with DIN 12576
  • Popular water-jacketed condenser suitable for distillation and reflux applications
  • With detachable plastic screwthread connectors for easy and safe attachment of rubber hose
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DIN 12576
380 mm
Borosilicate Glass
Suitable for distillation and reflux applications
19/26
Liebig Condenser
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Quickfit™ Borosilicate Glass Liebig Condenser > Cone Size: 19/26; Length: 380mm; Effective Length: 250mm