Learn More
Quickfit™ Borosilicate Glass Short Neck Round Bottom Flask
Use these flasks for a variety of applications. Quickfit™ Borosilicate Glass Short Neck Round Bottom Flask are manufactured from borosilicate glass for corrosion-free performance and resistance to thermal shock.
Brand:  Quickfit FR5/1S
 View more versions of this product
Product Code. 10407430
  • £153.00 / Pack of 5
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Use these flasks for a variety of applications. Quickfit™ Borosilicate Glass Short Neck Round Bottom Flask are manufactured from borosilicate glass for corrosion-free performance and resistance to thermal shock.
  • With highest quality Quickfit™ ground glass socket
  • Manufactured from borosilicate glass for corrosion free performance and resistance to thermal shock
  • Comprehensive range of 46 flasks from 5mL to 10,000ml
  • Body diameter and wall thickness comply with relevant ISO 1773 and ISO 4797 requirements
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 1773, ISO 4797
Borosilicate Glass
Short
Round
5 mL
65 mm
14/23
25 mm