Learn More
Quickfit™ Borosilicate Glass Single Neck Pear Shaped Flask
Allows distillation down to small quantities.
Brand:  Quickfit FP10/1
20.38 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 12020573
  • £22.00 / Each
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Allows distillation down to small quantities.
  • With highest quality Quickfit™ ground glass socket
  • Ideal for semi-micro distillations
  • Pear shaped design allows distillation down to small quantities with minimum hold-up, and reduced losses from thermal decomposition
  • Borosilicate glass for chemical resistance and thermal performance
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pear
Borosilicate Glass
28 mm
10 mL
85 mm