Learn More
Quickfit™ Borosilicate Glass Single Neck Pear Shaped Flask
Allows distillation down to small quantities.
£22.00 - £138.00
Specifications
Description Pear
Material Borosilicate Glass
View More Specs
Products 9
Product Code Brand Capacity (Metric) Diameter (Metric) Outer Height (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Diameter (Metric) Outer Height (Metric) Price Quantity & Availability  
11912088
View Documents Promotion Details
Quickfit
FP100/1
100 mL 58 mm 135 mm
£27.45
Each
Estimated Shipment: 15-01-2025
to see stock.
Add to basket
12030573
View Documents Promotion Details
Quickfit
FP100/2
100 mL 58 mm 140 mm
£27.45
Each
Estimated Shipment: 15-01-2025
to see stock.
Add to basket
12020573
View Documents Promotion Details
Quickfit
FP10/1
10 mL 28 mm 85 mm
£22.00
Each
Estimated Shipment: 28-11-2024
to see stock.
Add to basket
12090573
View Documents Promotion Details
Quickfit
FP25/1
25 mL 38 mm 100 mm
£24.90
Each
Estimated Shipment: 28-11-2024
to see stock.
Add to basket
12040603
View Documents Promotion Details
Quickfit
FP50/1
50 mL 48 mm 115 mm
£48.80
Each
Estimated Shipment: 12-12-2024
to see stock.
Add to basket
12060603
View Documents Promotion Details
Quickfit
FP50/2
50 mL 48 mm 125 mm
£24.65
Each
Estimated Shipment: 12-02-2025
to see stock.
Add to basket
15537850
View Documents Promotion Details
Quickfit
FP100/3
100 mL 58 mm 150 mm
£138.00
Pack of 5
Estimated Shipment: 15-01-2025
to see stock.
Add to basket
15547850
View Documents Promotion Details
Quickfit
FP25/0
25 mL 38 mm 89 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15567850
View Documents Promotion Details
Quickfit
FP5/1
5 mL 23 mm 67 mm
£102.00
Pack of 5
Estimated Shipment: 03-03-2025
to see stock.
Add to basket
Description
Description
Allows distillation down to small quantities.
  • With highest quality Quickfit™ ground glass socket
  • Ideal for semi-micro distillations
  • Pear shaped design allows distillation down to small quantities with minimum hold-up, and reduced losses from thermal decomposition
  • Borosilicate glass for chemical resistance and thermal performance
Specifications
Specifications
Pear
Borosilicate Glass