Learn More
Quickfit™ Three-Neck Round-Bottom Glass Flask
Useful for introducing gas inlet tubes and thermometers into the flask. The Quickfit Jointed Ware™ Quickfit™ Three-Neck Round-Bottom Flask has precision-ground sockets from borosilicate glass.
Brand:  Quickfit FR250/3S/22A
 View more versions of this product
Product Code. 12050653
  • £175.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Useful for introducing gas inlet tubes and thermometers into the flask. The Quickfit Jointed Ware™ Quickfit™ Three-Neck Round-Bottom Flask has precision-ground sockets from borosilicate glass.
  • Fitted with three Quickfit™ precision-ground sockets - One vertical centre neck - Two angled side necks
  • Quickfit™ ground glass sockets are manufactured to an accuracy which surpasses the requirements of ISO 383
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 383
Borosilicate Glass
1
Round
250 mL
130 mm
24/29 (Centre), 19/26 (side)