missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Quickfit™ Glass Key and Stem with Cone

Add liquid to most Quickfit™ flasks and assemblies. Pyrex™ Quickfit™ Glass Key and Stem with Cone includes stopcock with interchangeable glass key.

Brand:  Pyrex™ D1/22

101.48 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11351875

  • £198.00 / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Complies with ISO 4800
  • Funnel stem with ground glass cone for easy incorporation into Quickfit™ flasks and assemblies
  • Stopcock has interchangeable glass key
TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 mL
ISO 4800
Ground glass cone, stopcock with glass key
Dropping Funnel
Borosilicate Glass
19/26
2.5mm bore
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.