Learn More
Quickfit™ Funnel with Ground Glass Cone
Manufactured from chemically resistant Pyrex™ Borosilicate Glass. Quickfit™ Funnel with Ground Glass Cone is designed for transferring powders into Quickfit™ flasks and vessels.
Brand:  Quickfit CF5/5
51.69 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 11338054
  • £98.00 / Each
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Manufactured from chemically resistant Pyrex™ Borosilicate Glass. Quickfit™ Funnel with Ground Glass Cone is designed for transferring powders into Quickfit™ flasks and vessels.
  • Designed for transferring powders into Quickfit™ flasks and vessels
  • With Quickfit™ ground glass cone
  • Manufactured from chemically resistant Pyrex™ Borosilicate Glass
TRUSTED_SUSTAINABILITY
Specifications
Specifications
105mm
34/35
Powder Funnel
Borosilicate Glass
Quickfit™ Ground Glass Cone