Learn More
Quickfit™ Organic Chemistry Set
Starter kit - ideal for introducing students to principal techniques of preparative organic chemistry
Brand: Quickfit 29BU/M
170.46 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Starter kit - ideal for introducing students to principal techniques of preparative organic chemistry
- Comprises 5 glassware items all with size 14/23 Quickfit™ ground glass joints - giving maximum flexibility
- Scale based on starting material quantities up to 30g
- Both reflux and distillation assemblies can be constructed
- Housed in a robust cardboard box with foam rubber liner for product protection
Specifications
Organic chemistry kit | |
Five items of Glassware includes 50mL pear shaped flask, two still heads, three Liebig condenser, four screw cap adapters, five receiver adapter | |
Ideal for introducing students to the main techniques of preparative organic chemistry |