missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Reaction Vessel Bubbler with Inlet

Ideal for closing reaction mixture from the atmosphere

Brand:  Quickfit GB2/24SC

830.50 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15788017

  • £1185.00 / Pack of 5
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With gas inlet to provide continuous flow of inert gas (N2 or Ar) over the reaction
  • Complete with detachable plastic hose connection to allow easy and safe attachment to 9mm flexible tubing
  • Bubblers have capacity of approx 5mL
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Reaction Vessel Bubbler with Inlet
24/29
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers