missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Silicone Rings for Screw Thread Glass Joints

For use with Quickfit™ glass screw threads

Brand:  Quickfit QR24/9

Code : 01

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12989273

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

For use with Quickfit™ glass screwthreads

  • For use with Quickfit™ glass screwthreads
  • Melamine screwcaps with aperture at the top
  • Silicone rubber rings with PTFE washers
To determine what components are required: i) Check the size of the glass screwthread (size designation refers to O.D. of thread in mm) ii) Check the O.D. of the sampling tube, thermometer, probe etc. that is to be passed through the screwcap iii) Remember to order relevant cap, washer and ring separately
Specifications

Specifications

Rubber ring
Silicone
8.5 to 9.5mm
Thread size: 24mm
Quickfit™ glass screw threads
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers