Learn More
Quickfit™ Silicone Rings for Screw Thread Glass Joints
For use with Quickfit™ glass screw threads
£5.45 - £82.55
Specifications
Material Silicone
View More Specs
Products 9
Product Code Brand Diameter (Metric) Hole Size Price Quantity & Availability  
Product Code Brand Diameter (Metric) Hole Size Price Quantity & Availability  
12919283
View Documents Promotion Details
Quickfit
QR28/11
10.5 to 11.5 mm Thread size: 28mm
£82.55
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 23-10-2024
to see stock.
Add to basket
12969273
View Documents Promotion Details
Quickfit
QR18/7
7 to 8.5 mm Thread size: 18mm
£52.06
Pack of 10
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
12959273
View Documents Promotion Details
Quickfit
QR13/6
6 to 7mm Thread size: 13mm
£33.28
Pack of 10
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
12999273
View Documents Promotion Details
Quickfit
QR24/11
10.5 to 11.5 mm Thread size: 24mm
£44.89
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 23-10-2024
to see stock.
Add to basket
12979273
View Documents Promotion Details
Quickfit
QR24/6
6 to 7mm Thread size: 24mm
£47.47
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 23-10-2024
to see stock.
Add to basket
11302085
View Documents Promotion Details
Quickfit
QR28/6
6 to 7mm Thread size: 28mm
£71.15
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 13-09-2024
to see stock.
Add to basket
11392075
View Documents Promotion Details
Quickfit
QR28/18
18.0 to 19.0mm Thread size: 28mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15187193
View Documents Promotion Details
Quickfit
QR28/6
6 to 7mm Thread size: 28mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12989273
View Documents Promotion Details
Quickfit
QR24/9
8.5 to 9.5mm Thread size: 24mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
For use with Quickfit™ glass screwthreads
  • For use with Quickfit™ glass screwthreads
  • Melamine screwcaps with aperture at the top
  • Silicone rubber rings with PTFE washers
To determine what components are required: i) Check the size of the glass screwthread (size designation refers to O.D. of thread in mm) ii) Check the O.D. of the sampling tube, thermometer, probe etc. that is to be passed through the screwcap iii) Remember to order relevant cap, washer and ring separately
Specifications
Specifications
Silicone