missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Quiltec™ I Wipes
GREENER_CHOICEBIOPHARMA

Clean interior surfaces, tools, or products. Quiltec™ I Wipes offer an extremely high sorptive capacity. These durable polyester wipes are ideal for cleanroom applications.

Brand:  Contec™ QT1-99B

391.00 GBP valid until 2024-12-31
Use promo code "21919" to get your promotional price.


 View more versions of this product

Product Code. 13516940

  • £507.00 / Pack of 900
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Clean interior surfaces, tools, or products. Quiltec™ I Wipes offer an extremely high sorptive capacity. These durable polyester wipes are ideal for cleanroom applications.

  • These sealed-edge wipes offer an extremely high sorptive capacity. Two layers of quilted polyester fabric are laser-cut and sealed together to create pockets where additional fluid can be trapped.
  • Two-ply quilted polyester construction creates durable wiper with exceptional absorbency—absorbs more than twice the capacity of a single-layer wiper
  • Extremely low in both particles and fibers—ideal for interior surface cleaning of process tools, wiping down products, or general-purpose cleaning

Order Info

Laundered and packaged in an ISO Class 4 cleanroom.

Specifications

Specifications

Cleanroom Wipe
23 cm
23 cm
Not Autoclavable
Single-use
Cleanroom
2-ply
9 in.
9 in.
Polyester
ISO Class 3 and 4
23 x 23 cm (9 x 9 in.)
150/Bag, 6 Bags/Cs.
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers