Learn More
Thermo Scientific Chemicals Quinoline Yellow, Water Soluble 95%
£84.40 - £258.00
Chemical Identifiers
CAS | 8004-92-0 |
---|---|
Molecular Formula | C18H9NNa2O8S2 |
Molecular Weight (g/mol) | 477.37 |
MDL Number | MFCD00080727 |
InChI Key | FZUOVNMHEAPVBW-UHFFFAOYSA-L |
PubChem CID | 129893443 |
SMILES | [Na+].[Na+].[O-]S(=O)(=O)C1=CC(=C2N=C(C=CC2=C1)C1C(=O)C2=CC=CC=C2C1=O)S([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10063400
|
Thermo Scientific Chemicals
277340250 |
25 g | Glass bottle |
£84.40
25g |
In Stock
Log in to see stock. |
||||
11986921
|
Thermo Scientific Chemicals
277341000 |
100 g | Glass bottle |
£258.00
100g |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
8004-92-0 | |
477.37 | |
FZUOVNMHEAPVBW-UHFFFAOYSA-L | |
[Na+].[Na+].[O-]S(=O)(=O)C1=CC(=C2N=C(C=CC2=C1)C1C(=O)C2=CC=CC=C2C1=O)S([O-])(=O)=O |
C18H9NNa2O8S2 | |
MFCD00080727 | |
129893443 |
Specifications
8004-92-0 | |
C18H9NNa2O8S2 | |
Solubility in water: 225g/L (20°C). | |
Authentic | |
disodium 2-(1,3-dioxo-2,3-dihydro-1H-inden-2-yl)quinoline-6,8-disulfonate | |
129893443 | |
Pure | |
Quinoline Yellow |
95% | |
MFCD00080727 | |
FZUOVNMHEAPVBW-UHFFFAOYSA-L | |
[Na+].[Na+].[O-]S(=O)(=O)C1=CC(=C2N=C(C=CC2=C1)C1C(=O)C2=CC=CC=C2C1=O)S([O-])(=O)=O | |
477.37 | |
95% | |
Powder |
Safety and Handling
GHS P Statement
Harmful if swallowed.
GHS P Statement
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
- Quinoline Yellow, water soluble
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
RUO – Research Use Only