Learn More
(R)-(-)-2-Amino-3-methylbutane, ChiPros 98%, ee 97%, Thermo Scientific™
£54.80 - £162.00
Chemical Identifiers
CAS | 34701-33-2 |
---|---|
Molecular Formula | C5H13N |
Molecular Weight (g/mol) | 87.166 |
MDL Number | MFCD01075731 |
InChI Key | JOZZAIIGWFLONA-RXMQYKEDSA-N |
Synonym | r---2-amino-3-methylbutane, r---3-methyl-2-butylamine, r-3-methyl-2-butylamine, r-3-methylbutan-2-amine, 2r-3-methylbutan-2-amine, unii-s9ygl88qkq, s9ygl88qkq, 2r-3-methyl-butan-2-amine, 2-butanamine, 3-methyl-, r, r-3-methyl-2-butanamine |
PubChem CID | 6999786 |
IUPAC Name | (2R)-3-methylbutan-2-amine |
SMILES | CC(C)C(C)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11339105
|
Thermo Scientific Alfa Aesar
L16330.03 |
1 g |
£54.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11349105
|
Thermo Scientific Alfa Aesar
L16330.06 |
5 g |
£162.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications(R)-(-)-2-Amino-3-methylbutane is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in water (60g/L at 20°C).
Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store under dry inert gas. Incompatible with oxidizing agents. Store in cool place. It is sensitive to air.
Chemical Identifiers
34701-33-2 | |
87.166 | |
JOZZAIIGWFLONA-RXMQYKEDSA-N | |
6999786 | |
CC(C)C(C)N |
C5H13N | |
MFCD01075731 | |
r---2-amino-3-methylbutane, r---3-methyl-2-butylamine, r-3-methyl-2-butylamine, r-3-methylbutan-2-amine, 2r-3-methylbutan-2-amine, unii-s9ygl88qkq, s9ygl88qkq, 2r-3-methyl-butan-2-amine, 2-butanamine, 3-methyl-, r, r-3-methyl-2-butanamine | |
(2R)-3-methylbutan-2-amine |
Specifications
34701-33-2 | |
85°C to 87°C | |
C5H13N | |
MFCD01075731 | |
1718798 | |
r---2-amino-3-methylbutane, r---3-methyl-2-butylamine, r-3-methyl-2-butylamine, r-3-methylbutan-2-amine, 2r-3-methylbutan-2-amine, unii-s9ygl88qkq, s9ygl88qkq, 2r-3-methyl-butan-2-amine, 2-butanamine, 3-methyl-, r, r-3-methyl-2-butanamine | |
JOZZAIIGWFLONA-RXMQYKEDSA-N | |
CC(C)C(C)N | |
87.166 | |
87.16 | |
ee 97% |
0.746 | |
<21°C (69°F) | |
1.406 | |
UN2733 | |
Air Sensitive | |
Soluble in water (60g/L at 20°C). | |
−5.2° (Neat) | |
(2R)-3-methylbutan-2-amine | |
6999786 | |
98% | |
(R)-(-)-2-Amino-3-methylbutane |
Safety and Handling
GHS H Statement
H225-H311-H314-H318-H302-H332
Highly flammable liquid and vapor.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302-H314-H331-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 628-450-9
missing translation for 'rtecsNumber' : UI1100000
missing translation for 'tsca' : No
Recommended Storage : Store under Argon