missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Aminononane, ChiPros 99+%, ee 98+%, Thermo Scientific Chemicals

£25.90 - £96.70

Chemical Identifiers

CAS 74069-74-2
Molecular Formula C9H21N
Molecular Weight (g/mol) 143.274
MDL Number MFCD03844739
InChI Key ALXIFCUEJWCQQL-SECBINFHSA-N
Synonym r-2-aminononane, r---2-aminononane, 2r-nonan-2-amine, r-2-nonanamine, r-2-nonylamine, 2-nonanamine, 2r, r-2-aminononane, chipros r , produced by basf
PubChem CID 22831496
IUPAC Name (2R)-nonan-2-amine
SMILES CCCCCCCC(C)N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11394216
View Documents
Thermo Scientific Alfa Aesar
L19310.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11304226
View Documents
Thermo Scientific Alfa Aesar
L19310.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

74069-74-2
143.274
ALXIFCUEJWCQQL-SECBINFHSA-N
22831496
CCCCCCCC(C)N
C9H21N
MFCD03844739
r-2-aminononane, r---2-aminononane, 2r-nonan-2-amine, r-2-nonanamine, r-2-nonylamine, 2-nonanamine, 2r, r-2-aminononane, chipros r , produced by basf
(2R)-nonan-2-amine

Specifications

74069-74-2
73°C (19 mmHg)
C9H21N
MFCD03844739
Air Sensitive
ALXIFCUEJWCQQL-SECBINFHSA-N
CCCCCCCC(C)N
143.274
143.27
ee ≥98%
0.782
71°C (159°F)
1.4271
UN2735
r-2-aminononane, r---2-aminononane, 2r-nonan-2-amine, r-2-nonanamine, r-2-nonylamine, 2-nonanamine, 2r, r-2-aminononane, chipros r , produced by basf
−6° (Neat)
(2R)-nonan-2-amine
22831496
≥99%
(R)-(-)-2-Aminononane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 663-738-8

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only