missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2-(BOC-Amino)-1,4-butanediol, 97%, Thermo Scientific Chemicals

£50.10 - £151.00

Chemical Identifiers

CAS 397246-14-9
Molecular Formula C9H19NO4
Molecular Weight (g/mol) 205.25
InChI Key KLRRFBSWOIUAHZ-SSDOTTSWSA-N
Synonym r-+-2-boc-amino-1,4-butanediol, r-tert-butyl 1,4-dihydroxybutan-2-yl carbamate, r-2-boc-amino-butane-1,4-diol, r-2-boc-amino-1,4-butanediol, tert-butyl n-2r-1,4-dihydroxybutan-2-yl carbamate, pubchem15259, ksc498c9f, 1r-3-hydroxy-1-hydroxymethyl propyl carbamic acid tert-butyl ester, r-2-boc-amino-butan-1,4-diol, 2r-tert-butoxycarbonylamino butane-1,4-diol
PubChem CID 11954482
IUPAC Name tert-butyl N-[(2R)-1,4-dihydroxybutan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)NC(CCO)CO
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11935431
View Documents
Thermo Scientific Acros
435570010
1 g
£50.10
1g
Please to purchase this item. Need a web account? Register with us today!
11945431
View Documents
Thermo Scientific Acros
435570050
5 g
£151.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

397246-14-9
205.25
r-+-2-boc-amino-1,4-butanediol, r-tert-butyl 1,4-dihydroxybutan-2-yl carbamate, r-2-boc-amino-butane-1,4-diol, r-2-boc-amino-1,4-butanediol, tert-butyl n-2r-1,4-dihydroxybutan-2-yl carbamate, pubchem15259, ksc498c9f, 1r-3-hydroxy-1-hydroxymethyl propyl carbamic acid tert-butyl ester, r-2-boc-amino-butan-1,4-diol, 2r-tert-butoxycarbonylamino butane-1,4-diol
tert-butyl N-[(2R)-1,4-dihydroxybutan-2-yl]carbamate
C9H19NO4
KLRRFBSWOIUAHZ-SSDOTTSWSA-N
11954482
CC(C)(C)OC(=O)NC(CCO)CO

Specifications

66.4°C to 72.1°C
96.0
White
96% min. (GC)
7.8
KLRRFBSWOIUAHZ-SSDOTTSWSA-N
tert-butyl N-[(2R)-1,4-dihydroxybutan-2-yl]carbamate
11954482
97%
(R)-(+)-2-(BOC-Amino)-1, 4-butanediol
397246-14-9
100.0
Authentic
C9H19NO4
r-+-2-boc-amino-1,4-butanediol, r-tert-butyl 1,4-dihydroxybutan-2-yl carbamate, r-2-boc-amino-butane-1,4-diol, r-2-boc-amino-1,4-butanediol, tert-butyl n-2r-1,4-dihydroxybutan-2-yl carbamate, pubchem15259, ksc498c9f, 1r-3-hydroxy-1-hydroxymethyl propyl carbamic acid tert-butyl ester, r-2-boc-amino-butan-1,4-diol, 2r-tert-butoxycarbonylamino butane-1,4-diol
CC(C)(C)OC(=O)NC(CCO)CO
205.25
205.25
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

Product Identifier
  • (R)-(+)-2-(BOC-Amino)-1,4-butanediol

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only