missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Methylpiperazine, 98+%, Thermo Scientific Chemicals

£18.10 - £130.00

Chemical Identifiers

CAS 75336-86-6
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.165
MDL Number MFCD00192317
InChI Key JOMNTHCQHJPVAZ-RXMQYKEDSA-N
Synonym r---2-methylpiperazine, r-2-methylpiperazine, 2r-2-methylpiperazine, r-mp, piperazine, 2-methyl-, 2r, r-2-methyl-piperazine, 2-methylpiperazine, r, unii-p4xu643r8w
PubChem CID 7330434
IUPAC Name (2R)-2-methylpiperazine
SMILES CC1CNCCN1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11446106
View Documents
Thermo Scientific Alfa Aesar
B22366.01
0.25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11426106
View Documents
Thermo Scientific Alfa Aesar
B22366.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11436106
View Documents
Thermo Scientific Alfa Aesar
B22366.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

75336-86-6
100.165
JOMNTHCQHJPVAZ-RXMQYKEDSA-N
7330434
CC1CNCCN1
C5H12N2
MFCD00192317
r---2-methylpiperazine, r-2-methylpiperazine, 2r-2-methylpiperazine, r-mp, piperazine, 2-methyl-, 2r, r-2-methyl-piperazine, 2-methylpiperazine, r, unii-p4xu643r8w
(2R)-2-methylpiperazine

Specifications

75336-86-6
155°C to 156°C
C5H12N2
UN2925
Air sensitive; Hygroscopic
JOMNTHCQHJPVAZ-RXMQYKEDSA-N
CC1CNCCN1
100.165
100.17
(R)-(-)-2-Methylpiperazine
76°C to 78°C
65°C (149°F)
MFCD00192317
3647994
r---2-methylpiperazine, r-2-methylpiperazine, 2r-2-methylpiperazine, r-mp, piperazine, 2-methyl-, 2r, r-2-methyl-piperazine, 2-methylpiperazine, r, unii-p4xu643r8w
−7° (c=1 in Ethanol)
(2R)-2-methylpiperazine
7330434
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.

P210-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H228-H314

missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: II

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only