missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-4-Methylmandelic acid, ChiPros, 98%, ee 97+%, Thermo Scientific Chemicals

£50.10 - £552.75

Chemical Identifiers

CAS 31284-89-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00798433,MFCD00961860
InChI Key SFGURAWGCAPHON-UHFFFAOYNA-N
Synonym 2-hydroxy-2-4-methylphenyl acetic acid, 4-methylmandelic acid, p-methyl mandelic acid, 2-hydroxy-2-p-tolyl acetic acid, hydroxy 4-methylphenyl acetic acid, hydroxy-p-tolyl-acetic acid, p-methylmandelic acid, 4-methylmaldelic acid, p-methyl-mandelic acid, acmc-1afsj
PubChem CID 12458823
IUPAC Name (2R)-2-hydroxy-2-(4-methylphenyl)acetic acid
SMILES CC1=CC=C(C=C1)C(O)C(O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11405730
View Documents
Thermo Scientific Alfa Aesar
H25953.03
1 g
£50.10
1g
Please to purchase this item. Need a web account? Register with us today!
11415730
View Documents
Thermo Scientific Alfa Aesar
H25953.06
5 g
£161.00
5g
Please to purchase this item. Need a web account? Register with us today!
11425730
View Documents
Thermo Scientific Alfa Aesar
H25953.14
25 g
£552.75
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

31284-89-6
166.18
SFGURAWGCAPHON-UHFFFAOYNA-N
12458823
CC1=CC=C(C=C1)C(O)C(O)=O
C9H10O3
MFCD00798433,MFCD00961860
2-hydroxy-2-4-methylphenyl acetic acid, 4-methylmandelic acid, p-methyl mandelic acid, 2-hydroxy-2-p-tolyl acetic acid, hydroxy 4-methylphenyl acetic acid, hydroxy-p-tolyl-acetic acid, p-methylmandelic acid, 4-methylmaldelic acid, p-methyl-mandelic acid, acmc-1afsj
(2R)-2-hydroxy-2-(4-methylphenyl)acetic acid

Specifications

31284-89-6
C9H10O3
Light sensitive
SFGURAWGCAPHON-UHFFFAOYNA-N
CC1=CC=C(C=C1)C(O)C(O)=O
166.18
166.18
ee ≥97%
130°C to 133°C
MFCD00798433,MFCD00961860
2-hydroxy-2-4-methylphenyl acetic acid, 4-methylmandelic acid, p-methyl mandelic acid, 2-hydroxy-2-p-tolyl acetic acid, hydroxy 4-methylphenyl acetic acid, hydroxy-p-tolyl-acetic acid, p-methylmandelic acid, 4-methylmaldelic acid, p-methyl-mandelic acid, acmc-1afsj
−154° (c=1 in Methanol)
(2R)-2-hydroxy-2-(4-methylphenyl)acetic acid
12458823
98%
(R)-(-)-4-Methylmandelic acid, ChiPros
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c

H315-H318-H335

missing translation for 'einecsNumber' : 662-983-8

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only