Learn More
R&D Systems™ Cultrex RGF Basement Membrane Extract with Phenol Red
soluble form of basement membrane purified from Engelbreth-Holm-Swarm (EHS) tumor
Brand: R&D Systems™ 3433-010-01P
Additional Details : Weight : 0.00970kg
Description
Functional Assay - Tube formation assay: BME promotes formation of capillary-like structures by human (HBMVEC; HUVEC) or mouse (SVEC4-10) endothelial cells. Gelling Assay - BME gels in less than 30 minutes at 37degreesC, and maintains the gelled form in culture medium for a minimum of 14 days at 37degreesC.Specifications
Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles. | |
≤8EU/mL | |
2 x 5 mL |
12 to 18 mg/mL | |
Bioactivity | |
Murine Engelbreth-Holm-Swarm (EHS) tumor. |