missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Cultrex RGF Basement Membrane Extract with Phenol Red
soluble form of basement membrane purified from Engelbreth-Holm-Swarm (EHS) tumor
£248.37 - £387.68
Specifications
Content And Storage | Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles. |
---|---|
Concentration | 12 to 18 mg/mL |
Endotoxin Level | ≤8EU/mL |
For Use With (Application) | Bioactivity |
Source | Murine Engelbreth-Holm-Swarm (EHS) tumor. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18674759
|
R&D Systems™
3433-010-01P |
2 x 5 mL |
£387.68
10mL |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18412370
|
R&D Systems™
3433-005-01P |
5 mL |
£248.37
5mL |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The extract gels at 37°C to form a reconstituted basement membrane. The major components of BME include laminin, collagen IV, entactin, and heparin sulfate proteoglycan. BME can be used in a multiple applications, under a variety of cell culture conditions, for maintaining growth or promoting differentiation of primary endothelial, epithelial, smooth muscle and stem cells. BME can also be utilized in cell attachment, neurite outgrowth, angiogenesis, in vitro cell invasion and in vivo tumorigenicity assays.Specifications
Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles. | |
≤8EU/mL | |
Murine Engelbreth-Holm-Swarm (EHS) tumor. |
12 to 18 mg/mL | |
Bioactivity |