missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-5 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£216.00 - £616.00
Specifications
Gene ID (Entrez) | 3567 |
---|---|
Name | IL-5 |
Source | Spodoptera frugiperda,Sf 21 (stably transfected)-derived human IL-5 protein Ile20-Ser134 |
Structural Form | Disulfide-linked homodimer |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Formulation | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Formulation | Price | Quantity & Availability | ||||
18108540
|
R&D Systems™
205-IL-005/CF |
5 μg | Supplied as a 0.2μm filtered solution in PBS. |
£216.00
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18186690
|
R&D Systems™
205-IL-025 |
25 μg | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£616.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18197780
|
R&D Systems™
205-IL-025/CF |
25 μg | Supplied as a 0.2μm filtered solution in PBS. |
£616.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18147250
|
R&D Systems™
205-IL-005 |
5 μg | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£216.00
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Human IL-5 Protein is derived from Sf 21 (stably transfected). The Recombinant Human IL-5 Protein has been validated for the following applications: Bioactivity.Specifications
3567 | |
Spodoptera frugiperda,Sf 21 (stably transfected)-derived human IL-5 protein Ile20-Ser134 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
IL-5 | |
Disulfide-linked homodimer | |
Recombinant |