Learn More
R&D Systems™ Mouse Erythrocyte Lysing Kit
For the removal of erythrocytes (RBC) from splenocyte preparations
Brand: R&D Systems™ WL2000
Additional Details : Weight : 0.00970kg
Description
- Gently lyses erythrocytes
- Stabilizes leukocytes for analysis by flow cytometry
- Maintains natural light scattering and fluorescent staining properties of leukocytes
- Maintains viability of leukocytes for subsequent tissue culture manipulation
- Gently lyses erythrocytes
- Stabilizes leukocytes for analysis by flow cytometry
- Maintains natural light scattering and fluorescent staining properties of leukocytes
- Maintains viability of leukocytes for subsequent tissue culture manipulation
Specifications
M-Lyse Buffer Concentrate (10X); Wash Buffer Concentrate (10X); Fixative Concentrate (10X) | |
Store at 20°C to 25°C | |
12.5 x 109 splenocytes |
Cell Lysis Kit | |
Removal of erythrocytes (red blood cells) from whole blood |